Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYOU1 | NM_001130991.2 | NP_001124463.1 | ||
| NM_006389.4 | NP_006380.1 | |||
| XM_005271392.3 | XP_005271449.1 | |||
| XM_005271393.3 | XP_005271450.1 | |||
| XM_005271394.3 | XP_005271451.1 | |||
| XM_017017095.1 | XP_016872584.1 | |||
| XM_017017096.1 | XP_016872585.1 | |||
| XM_017017097.1 | XP_016872586.1 | |||
| AK127324.1 | ||||
| AK222756.1 | BAD96476.1 | |||
| AK294905.1 | ||||
| AK297260.1 | ||||
| AK299648.1 | ||||
| AK301514.1 | ||||
| AK304264.1 | ||||
| AK307624.1 | ||||
| AK314178.1 | ||||
| BC004560.2 | AAH04560.1 | |||
| BC017726.1 | ||||
| BC072436.1 | AAH72436.1 | |||
| DQ350134.1 | ABC75106.1 | |||
| DQ372932.1 | ABD14370.1 | |||
| U65785.1 | AAC50947.1 |
More Information
Set Membership: |
|

Genomic Map