Assay Details
Target Gene Details
Entrez Gene ID: | 25913 |
Gene Name: | protection of telomeres 1 |
Gene Aliases: |
CMM10, GLM9, HPOT1 |
Location: |
Chr.7:124822386-124929983 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POT1 | NM_001042594.1 | NP_001036059.1 | ||
| NM_015450.2 | NP_056265.2 | |||
| NR_003102.1 | ||||
| NR_003103.1 | ||||
| NR_003104.1 | ||||
| XM_006715917.3 | XP_006715980.1 | |||
| XM_011516006.1 | XP_011514308.1 | |||
| XM_011516007.1 | XP_011514309.1 | |||
| XM_017011942.1 | XP_016867431.1 | |||
| AK001230.1 | BAA91568.1 | |||
| AK001935.1 | BAA91988.1 | |||
| AK022580.1 | BAB14110.1 | |||
| AK026234.1 | BAB15404.1 | |||
| AK302263.1 | ||||
| AL050120.1 | CAB43281.1 | |||
| AY823520.1 | AAW22613.1 | |||
| AY823521.1 | AAW22614.1 | |||
| AY823522.1 | AAW22615.1 | |||
| AY823524.1 | AAW22617.1 | |||
| BC002923.2 | AAH02923.1 | |||
| BM693856.1 |
More Information
Set Membership: |
|

Genomic Map