Assay Details
Target Gene Details
Entrez Gene ID: | 389677 |
Gene Name: | RNA binding motif protein 12B |
Gene Aliases: |
MGC:33837 |
Location: |
Chr.8:93725161-93741012 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM12B | NM_203390.2 | 3 | 3616 | NP_976324.2 |
| XM_005250915.4 | 4 | 3693 | XP_005250972.1 | |
| XM_011517027.2 | 3 | 4342 | XP_011515329.1 | |
| XM_011517028.2 | 3 | 3637 | XP_011515330.1 | |
| XM_011517029.2 | 2 | 3583 | XP_011515331.1 | |
| XM_017013406.1 | 3 | 3876 | XP_016868895.1 | |
| XM_017013407.1 | 5 | 3748 | XP_016868896.1 | |
| XM_017013408.1 | 3 | 3636 | XP_016868897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831399 | Chr.8:93703175 - 93892637 on Build GRCh38 | Loss |
|
| nsv6309 | Chr.8:93729557 - 93763774 on Build GRCh38 | Insertion |
|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map