Assay Details
Target Gene Details
Entrez Gene ID: | 7022 |
Gene Name: | transcription factor AP-2 gamma |
Gene Aliases: |
AP2-GAMMA, ERF1, TFAP2G, hAP-2g |
Location: |
Chr.20:56629302-56639283 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFAP2C | NM_003222.3 | 4 | 954 | NP_003213.1 |
| AK301572.1 | 4 | 770 | ||
| AK314272.1 | 4 | 950 | ||
| BC035664.1 | 4 | 913 | AAH35664.2 | |
| BC051829.1 | 4 | 954 | AAH51829.2 | |
| U85658.1 | 4 | 877 | AAC51305.1 | |
| X95693.1 | 4 | 757 | CAA64989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| nsv1061975 | Chr.20:56580562 - 57136322 on Build GRCh38 | Gain |
|
| nsv834015 | Chr.20:56518712 - 56656502 on Build GRCh38 | Loss |
|
| nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map