Assay Details
Target Gene Details
Entrez Gene ID: | 7128 |
Gene Name: | TNF alpha induced protein 3 |
Gene Aliases: |
A20, AISBL, OTUD7C, TNFA1P2 |
Location: |
Chr.6:137866317-137883314 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP3 | NM_001270507.1 | 7 | 1764 | NP_001257436.1 |
| NM_001270508.1 | 7 | 1788 | NP_001257437.1 | |
| NM_006290.3 | 7 | 1740 | NP_006281.1 | |
| XM_005267119.1 | 7 | 1507 | XP_005267176.1 | |
| XM_006715555.1 | 5 | 1245 | XP_006715618.1 | |
| XM_011536095.1 | 8 | 1783 | XP_011534397.1 | |
| XM_011536096.1 | 7 | 1768 | XP_011534398.1 | |
| AK312862.1 | 7 | 1692 | ||
| AL157444.1 | 5 | 956 | CAB75664.1 | |
| BC113871.1 | 6 | 1441 | AAI13872.1 | |
| BC114480.1 | 6 | 1441 | AAI14481.1 | |
| FJ434405.1 | 6 | 1418 | ||
| FJ434408.1 | 6 | 1418 | ||
| FJ434409.1 | ||||
| FJ434411.1 | 6 | 1418 | ||
| FJ434412.1 | 6 | 1418 | ||
| FJ434413.1 | 6 | 1418 | ||
| FJ434414.1 | 6 | 1418 | ||
| M59465.1 | 7 | 1484 | AAA51550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830818 | Chr.6:137790698 - 137950293 on Build GRCh38 | Gain |
|
| nsv5493 | Chr.6:137849427 - 137883180 on Build GRCh38 | Insertion |
|
| nsv830819 | Chr.6:137841569 - 138013923 on Build GRCh38 | Loss |
|
| nsv470863 | Chr.6:137080640 - 138014159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map