Assay Details
Target Gene Details
Entrez Gene ID: | 341799 |
Gene Name: | olfactory receptor family 6 subfamily S member 1 |
Gene Aliases: |
OR14-37, OR6S1Q |
Location: |
Chr.14:20640696-20641691 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6S1 | NM_001001968.1 | 1 | 500 | NP_001001968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map