Assay Details
Target Gene Details
Entrez Gene ID: | 1509 |
Gene Name: | cathepsin D |
Gene Aliases: |
CLN10, CPSD, HEL-S-130P |
Location: |
Chr.11:1752752-1763992 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSD | NM_001909.4 | 7 | 1022 | NP_001900.1 |
| AK130178.1 | 7 | 914 | ||
| BC016320.2 | 7 | 934 | AAH16320.1 | |
| BT006910.1 | 7 | 889 | AAP35556.1 | |
| BT020155.1 | 7 | 889 | AAV38957.1 | |
| CR456947.1 | 7 | 889 | CAG33228.1 | |
| EU794598.1 | 7 | 934 | ||
| KU178007.1 | 7 | 889 | ||
| KU178008.1 | ||||
| M11233.1 | 7 | 940 | AAB59529.1 | |
| X05344.1 | 7 | 891 | CAA28955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832049 | Chr.11:1669923 - 1817378 on Build GRCh38 | Loss |
|
| dgv1564n54 | Chr.11:1752247 - 1785884 on Build GRCh38 | Loss |
|
| nsv7642 | Chr.11:1749402 - 1784428 on Build GRCh38 | Insertion |
|
| nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map