Assay Details
Target Gene Details
Entrez Gene ID: | 2121 |
Gene Name: | EvC ciliary complex subunit 1 |
Gene Aliases: |
DWF-1, EVC1, EVCL |
Location: |
Chr.4:5711197-5829043 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVC | NM_001306090.1 | NP_001293019.1 | ||
| NM_001306092.1 | NP_001293021.1 | |||
| NM_153717.2 | NP_714928.1 | |||
| XM_006713865.3 | XP_006713928.1 | |||
| XM_006713866.3 | XP_006713929.1 | |||
| XM_011513419.2 | XP_011511721.1 | |||
| XM_017007883.1 | XP_016863372.1 | |||
| AF216184.1 | AAF37216.1 | |||
| AF216185.1 | AAF37217.1 | |||
| AF239742.1 | AAF44682.1 | |||
| BC085608.1 | AAH85608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829844 | Chr.4:5719377 - 5925385 on Build GRCh38 | Gain |
|
| nsv508993 | Chr.4:5688142 - 5744093 on Build GRCh38 | Insertion |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map