Assay Details
Target Gene Details
Entrez Gene ID: | 23396 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 gamma |
Gene Aliases: |
LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma |
Location: |
Chr.19:3630181-3700492 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP5K1C | NM_001195733.1 | 17 | 4208 | NP_001182662.1 |
| NM_012398.2 | 18 | 4292 | NP_036530.1 | |
| XM_017026540.1 | 17 | 4266 | XP_016882029.1 | |
| AB011161.1 | 18 | 4260 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map