Assay Details
Target Gene Details
Entrez Gene ID: | 135293 |
Gene Name: | peptidase M20 domain containing 2 |
Gene Aliases: |
ACY1L2, bA63L7.3 |
Location: |
Chr.6:89096879-89165569 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PM20D2 | XM_011535481.2 | XP_011533783.1 |
Target Gene Details
Entrez Gene ID: | 135295 |
Gene Name: | serine and arginine rich splicing factor 12 |
Gene Aliases: |
SFRS13B, SFRS19, SRrp35 |
Location: |
Chr.6:89095957-89118081 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF12 | NM_080743.4 | NP_542781.3 | ||
| XM_006715348.2 | XP_006715411.1 | |||
| XM_011535483.2 | XP_011533785.1 | |||
| XM_017010292.1 | XP_016865781.1 | |||
| XM_017010293.1 | XP_016865782.1 | |||
| AF449428.1 | AAL57515.1 | |||
| AK314008.1 | ||||
| BC021715.1 | AAH21715.1 | |||
| BP244338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023532 | Chr.6:89093252 - 89253488 on Build GRCh38 | Loss |
|
| nsv1027007 | Chr.6:89099218 - 89628146 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map