Assay Details
Target Gene Details
Entrez Gene ID: | 1195 |
Gene Name: | CDC like kinase 1 |
Gene Aliases: |
CLK, CLK/STY, STY |
Location: |
Chr.2:200853009-200864744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK1 | NM_001162407.1 | NP_001155879.1 | ||
| NM_004071.3 | NP_004062.2 | |||
| NR_027855.1 | ||||
| NR_027856.1 | ||||
| AK025306.1 | ||||
| AK294295.1 | ||||
| AK316127.1 | ||||
| BC028149.1 | ||||
| BC031549.1 | AAH31549.1 | |||
| L29219.1 | AAA61480.1 | |||
| L29222.1 | AAB59459.1 | |||
| M59287.1 |
More Information
Set Membership: |
|

Genomic Map