Assay Details
Target Gene Details
Entrez Gene ID: | 100996928 |
Gene Name: | C7orf55-LUC7L2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:139341132-139423457 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf55-LUC7L2 | NM_001244584.2 | 10 | 1366 | NP_001231513.1 |
| AK297642.1 | 10 | 1138 |
Target Gene Details
Entrez Gene ID: | 100129148 |
Gene Name: | uncharacterized LOC100129148 |
Gene Aliases: |
- |
Location: |
Chr.7:139417462-139427526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129148 | NR_033999.1 | |||
| AI811258.1 |
Target Gene Details
Entrez Gene ID: | 51631 |
Gene Name: | LUC7 like 2, pre-mRNA splicing factor |
Gene Aliases: |
CGI-59, CGI-74, LUC7B2 |
Location: |
Chr.7:139340359-139423457 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUC7L2 | NM_001244585.1 | 10 | 1090 | NP_001231514.1 |
| NM_001270643.1 | 10 | 1402 | NP_001257572.1 | |
| NM_016019.4 | 9 | 1331 | NP_057103.2 | |
| AF151817.1 | 9 | 890 | AAD34054.1 | |
| AF151832.1 | 9 | 915 | AAD34069.1 | |
| AK001476.1 | 9 | 1223 | BAA91713.1 | |
| AK001519.1 | 9 | 1269 | BAA91737.1 | |
| AK022895.1 | 9 | 1206 | BAB14297.1 | |
| AK092044.1 | 8 | 1712 | ||
| AK093864.1 | 10 | 1189 | ||
| AK094081.1 | 1 | 1519 | ||
| AK298166.1 | 10 | 1090 | ||
| BC017163.2 | 9 | 1248 | AAH17163.1 | |
| BC042625.1 | 10 | 1336 | AAH42625.1 | |
| BC050708.2 | 9 | 1261 | AAH50708.1 | |
| BC056886.1 | 9 | 1206 | AAH56886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map