Assay Details
Target Gene Details
Entrez Gene ID: | 126308 |
Gene Name: | MOB kinase activator 3A |
Gene Aliases: |
MOB1C, MOBKL2A, moblak |
Location: |
Chr.19:2071036-2096502 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3A | NM_130807.2 | 3 | 307 | NP_570719.1 |
| XM_011527683.2 | 3 | 384 | XP_011525985.1 | |
| AB061793.1 | 2 | 170 | BAB84554.1 | |
| AJ580635.1 | 1 | 148 | CAE45267.1 | |
| AK095471.1 | 3 | 307 | ||
| AL832474.1 | 3 | 343 | ||
| BC015049.1 | 2 | 196 | AAH15049.1 | |
| CR627409.1 | 3 | 308 | CAH10498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss |
|
| nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss |
|
| esv3583203 | Chr.19:2044317 - 2080698 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map