Assay Details
Target Gene Details
Entrez Gene ID: | 23612 |
Gene Name: | pleckstrin homology like domain family A member 3 |
Gene Aliases: |
TIH1 |
Location: |
Chr.1:201464284-201469171 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLDA3 | NM_012396.4 | 2 | 901 | NP_036528.1 |
| NR_073080.1 | 3 | 995 | ||
| AF151100.1 | 2 | 835 | AAD42081.1 | |
| AK075179.1 | 2 | 864 | BAC11454.1 | |
| AK222644.1 | 2 | 864 | BAD96364.1 | |
| BC014390.1 | 2 | 753 | AAH14390.1 | |
| BC068273.1 | 2 | 901 | AAH68273.1 | |
| CN356697.1 | 3 | 407 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832315 | Chr.1:201364463 - 201525988 on Build GRCh38 | Loss |
|
| nsv470776 | Chr.1:201319827 - 201534604 on Build GRCh38 | Gain |
|
| nsv467961 | Chr.1:201316584 - 201534604 on Build GRCh38 | Gain |
|
| nsv832326 | Chr.1:201375720 - 201475887 on Build GRCh38 | Loss |
|
| nsv947466 | Chr.1:201308515 - 201540498 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map