Assay Details
Target Gene Details
Entrez Gene ID: | 9918 |
Gene Name: | non-SMC condensin I complex subunit D2 |
Gene Aliases: |
CAP-D2, CNAP1, hCAP-D2 |
Location: |
Chr.12:6494132-6531966 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPD2 | NM_014865.3 | 18 | 2315 | NP_055680.3 |
| AK022511.1 | 18 | 2315 | ||
| AK125155.1 | 15 | 1970 | BAC86066.1 | |
| AK128354.1 | 18 | 2227 | ||
| AK303310.1 | 17 | 2290 | ||
| BC028182.1 | 18 | 2327 | AAH28182.1 | |
| D63880.1 | 18 | 3068 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759876 | Chr.12:6344718 - 6554669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map