Assay Details
Target Gene Details
Entrez Gene ID: | 7572 |
Gene Name: | zinc finger protein 24 |
Gene Aliases: |
KOX17, RSG-A, ZNF191, ZSCAN3, Zfp191 |
Location: |
Chr.18:35332208-35344467 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF24 | NM_001308123.1 | NP_001295052.1 | ||
| NM_006965.3 | NP_008896.2 | |||
| XM_005258341.3 | XP_005258398.1 | |||
| AF038964.1 | AAD19827.1 | |||
| AF542097.1 | AAN40767.1 | |||
| AK291246.1 | ||||
| BC003566.1 | AAH03566.1 | |||
| BC016801.1 | AAH16801.1 | |||
| BG436261.1 | ||||
| BT006658.1 | AAP35304.1 | |||
| CA314100.1 | ||||
| CR450328.1 | CAG29324.1 | |||
| U68536.1 | AAB37275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059793 | Chr.18:35259520 - 35361982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map