Assay Details
Target Gene Details
Entrez Gene ID: | 9985 |
Gene Name: | REC8 meiotic recombination protein |
Gene Aliases: |
HR21spB, REC8L1, Rec8p |
Location: |
Chr.14:24171853-24180408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REC8 | NM_001048205.1 | NP_001041670.1 | ||
| NM_005132.2 | NP_005123.2 | |||
| XM_011537421.1 | XP_011535723.1 | |||
| XM_017021841.1 | XP_016877330.1 | |||
| AF006264.1 | AAD01193.1 | |||
| AK001435.1 | BAA91690.1 | |||
| AK291191.1 | ||||
| AL832973.1 | CAH56339.1 | |||
| BC004159.2 | AAH04159.1 | |||
| BC010887.1 | AAH10887.1 | |||
| CR457263.1 | CAG33544.1 | |||
| DB294585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522667 | Chr.14:24172317 - 24185384 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map