Assay Details
Target Gene Details
Entrez Gene ID: | 49860 |
Gene Name: | cornulin |
Gene Aliases: |
C1orf10, DRC1, PDRC1, SEP53 |
Location: |
Chr.1:152409243-152414274 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRNN | NM_016190.2 | 2 | 111 | NP_057274.1 |
| AF077831.1 | 2 | 89 | AAD55747.1 | |
| AK291301.1 | 2 | 100 | ||
| AK298640.1 | 2 | 100 | ||
| AK316568.1 | 2 | 100 | ||
| BC030807.1 | 2 | 100 | AAH30807.1 | |
| DA420219.1 | 2 | 111 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014095 | Chr.1:152132010 - 152482115 on Build GRCh38 | Gain |
|
| nsv516541 | Chr.1:152404234 - 152417976 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map