Assay Details
Target Gene Details
Entrez Gene ID: | 1026 |
Gene Name: | cyclin dependent kinase inhibitor 1A |
Gene Aliases: |
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1 |
Location: |
Chr.6:36676460-36687339 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN1A | NM_000389.4 | NP_000380.1 | ||
| NM_001220777.1 | NP_001207706.1 | |||
| NM_001220778.1 | 1 | 75 | NP_001207707.1 | |
| NM_001291549.1 | NP_001278478.1 | |||
| NM_078467.2 | NP_510867.1 | |||
| AB209881.1 | BAD93118.1 | |||
| AK298901.1 | ||||
| AK309512.1 | ||||
| BC000275.1 | AAH00275.1 | |||
| BC000312.2 | 1 | 59 | AAH00312.1 | |
| BC001935.1 | AAH01935.1 | |||
| BC013967.2 | AAH13967.1 | |||
| BU154329.1 | ||||
| BU193165.1 | ||||
| DA552678.1 | 1 | 75 | ||
| DA582789.1 | ||||
| DA699672.1 | ||||
| DA702185.1 | ||||
| DA838987.1 | ||||
| L25610.1 | AAA16109.1 | |||
| L26165.1 | AAA19811.1 | |||
| L47232.1 | AAB59559.1 | |||
| L47233.1 | AAB59560.1 | |||
| S67388.1 | AAB29246.1 | |||
| U03106.1 | AAC04313.1 | |||
| U09579.1 | AAA85641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map