Assay Details
Target Gene Details
Entrez Gene ID: | 127254 |
Gene Name: | glutamate rich 3 |
Gene Aliases: |
C1orf173 |
Location: |
Chr.1:74568111-74674343 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH3 | NM_001002912.4 | 10 | 1476 | NP_001002912.4 |
| XM_017000274.1 | 10 | 1476 | XP_016855763.1 | |
| XM_017000275.1 | 10 | 1266 | XP_016855764.1 | |
| AK127470.1 | 10 | 1476 | BAC86994.1 | |
| BC073916.1 | 5 | 844 | AAH73916.1 |
Target Gene Details
Entrez Gene ID: | 101927320 |
Gene Name: | ERICH3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:74577430-74626098 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH3-AS1 | NR_121670.1 | |||
| NR_121671.1 | ||||
| BX092187.1 | ||||
| CR627203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830248 | Chr.1:74496707 - 74678774 on Build GRCh38 | Loss |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|

Genomic Map