Assay Details
Target Gene Details
Entrez Gene ID: | 56917 |
Gene Name: | Meis homeobox 3 |
Gene Aliases: |
MRG2 |
Location: |
Chr.19:47403118-47422315 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEIS3 | NM_001009813.2 | NP_001009813.1 | ||
| NM_001301059.1 | NP_001287988.1 | |||
| NM_020160.2 | NP_064545.1 | |||
| XM_011527136.2 | XP_011525438.1 | |||
| XM_011527138.2 | XP_011525440.1 | |||
| XM_011527139.2 | XP_011525441.1 | |||
| XM_011527140.1 | XP_011525442.1 | |||
| XM_017027011.1 | XP_016882500.1 | |||
| AK098710.1 | ||||
| AK289950.1 | ||||
| AL537583.3 | ||||
| AY091593.1 | AAM09846.1 | |||
| BC025404.1 | AAH25404.1 | |||
| BC069251.1 | AAH69251.1 | |||
| BM783223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv517901 | Chr.19:47373953 - 47444971 on Build GRCh38 | Loss |
|
| nsv9738 | Chr.19:47356269 - 47427931 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map