Assay Details
Target Gene Details
Entrez Gene ID: | 116443 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 3A |
Gene Aliases: |
GluN3A, NMDAR-L, NR3A |
Location: |
Chr.9:101569352-101738580 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN3A | NM_133445.2 | 9 | 3873 | NP_597702.2 |
| AB075853.1 | 9 | 3873 | ||
| AF416558.1 | 9 | 3285 | AAL40734.1 | |
| AJ416950.2 | 9 | 3287 | CAC95229.2 | |
| AL359651.1 | 4 | 680 | ||
| BC132866.1 | 9 | 3699 | ||
| BC132868.1 | 9 | 3699 | ||
| BC167432.1 | 9 | 3873 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831672 | Chr.9:101538473 - 101718567 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map