Assay Details
Target Gene Details
Entrez Gene ID: | 3191 |
Gene Name: | heterogeneous nuclear ribonucleoprotein L |
Gene Aliases: |
HNRPL, P/OKcl.14, hnRNP-L |
Location: |
Chr.19:38836378-38852339 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPL | NM_001005335.1 | NP_001005335.1 | ||
| NM_001533.2 | NP_001524.2 | |||
| XM_011526886.1 | XP_011525188.1 | |||
| XM_011526887.1 | XP_011525189.1 | |||
| XM_011526888.2 | XP_011525190.1 | |||
| XM_011526889.1 | XP_011525191.1 | |||
| AB044547.1 | BAB18649.1 | |||
| AK092828.1 | ||||
| AK094966.1 | ||||
| AK097975.1 | ||||
| AK292115.1 | ||||
| AK313652.1 | ||||
| BC069184.1 | ||||
| BM997815.1 | ||||
| BQ334900.1 | ||||
| X16135.1 | CAA34261.1 |
More Information
Set Membership: |
|

Genomic Map