Assay Details
Target Gene Details
Entrez Gene ID: | 10979 |
Gene Name: | fermitin family member 2 |
Gene Aliases: |
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2 |
Location: |
Chr.14:52857271-52951192 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FERMT2 | NM_001134999.1 | 16 | 3239 | NP_001128471.1 |
| NM_006832.2 | 15 | 3218 | NP_006823.1 | |
| XM_005267285.2 | 16 | 3346 | XP_005267342.1 | |
| XM_006720008.2 | 18 | 3391 | XP_006720071.1 | |
| XM_006720009.2 | 17 | 3367 | XP_006720072.1 | |
| AF443279.1 | 15 | 3113 | AAN75823.1 | |
| AK126426.1 | 3 | 2268 | ||
| CR749486.1 | 2 | 5894 | ||
| Z24725.1 | 15 | 3154 | CAA80852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832796 | Chr.14:52680158 - 52860182 on Build GRCh38 | Loss |
|
| nsv1050582 | Chr.14:52773715 - 52861657 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map