Assay Details
Target Gene Details
Entrez Gene ID: | 83697 |
Gene Name: | solute carrier family 4 member 9 |
Gene Aliases: |
AE4 |
Location: |
Chr.5:140359645-140375177 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A9 | NM_001258426.1 | NP_001245355.1 | ||
| NM_001258427.1 | NP_001245356.1 | |||
| NM_001258428.1 | 2 | 359 | NP_001245357.1 | |
| NM_031467.2 | NP_113655.2 | |||
| XM_005268521.3 | XP_005268578.2 | |||
| XM_011537694.2 | 2 | 916 | XP_011535996.1 | |
| XM_017009933.1 | 2 | 916 | XP_016865422.1 | |
| XM_017009934.1 | 2 | 916 | XP_016865423.1 | |
| XM_017009935.1 | 2 | 916 | XP_016865424.1 | |
| XM_017009936.1 | XP_016865425.1 | |||
| XM_017009937.1 | 2 | 916 | XP_016865426.1 | |
| XM_017009938.1 | 2 | 916 | XP_016865427.1 | |
| XM_017009939.1 | XP_016865428.1 | |||
| XM_017009940.1 | 2 | 916 | XP_016865429.1 | |
| XM_017009941.1 | 2 | 916 | XP_016865430.1 | |
| XM_017009942.1 | 2 | 916 | XP_016865431.1 | |
| XM_017009943.1 | XP_016865432.1 | |||
| AB032762.1 | BAA93010.1 | |||
| AF313465.1 | 2 | 346 | AAK28832.1 | |
| AF332961.1 | AAK69625.1 | |||
| AF336237.1 | AAK16733.1 | |||
| BC136262.1 | ||||
| BC143602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509090 | Chr.5:140311306 - 140421024 on Build GRCh38 | Insertion |
|
| nsv1020011 | Chr.5:140340815 - 140396254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map