Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP44 | NM_001042403.2 | 2 | 1331 | NP_001035862.1 |
| NM_001278393.1 | 2 | 1547 | NP_001265322.1 | |
| NM_032147.4 | 2 | 1476 | NP_115523.2 | |
| NR_125360.1 | 1 | 1257 | ||
| XM_005269172.1 | 2 | 1470 | XP_005269229.1 | |
| XM_005269173.1 | 2 | 1547 | XP_005269230.1 | |
| XM_005269174.1 | 2 | 1314 | XP_005269231.1 | |
| XM_011538800.2 | 3 | 1497 | XP_011537102.1 | |
| XM_011538801.1 | 3 | 1357 | XP_011537103.1 | |
| XM_011538802.1 | 2 | 1317 | XP_011537104.1 | |
| XM_011538803.1 | 2 | 1322 | XP_011537105.1 | |
| XM_011538804.2 | 2 | 1530 | XP_011537106.1 | |
| XM_011538805.1 | 2 | 2171 | XP_011537107.1 | |
| XM_011538806.2 | 2 | 1470 | XP_011537108.1 | |
| XM_017020009.1 | 2 | 1465 | XP_016875498.1 | |
| XM_017020010.1 | 2 | 1317 | XP_016875499.1 | |
| XM_017020011.1 | 3 | 1343 | XP_016875500.1 | |
| XM_017020012.1 | 2 | 1470 | XP_016875501.1 | |
| AK027434.1 | 2 | 1282 | ||
| AK315697.1 | 2 | 1479 | ||
| AL136825.1 | 2 | 1437 | CAB66759.1 | |
| BC030704.1 | 2 | 1312 | AAH30704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052350 | Chr.12:95528912 - 95686094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map