Assay Details
Target Gene Details
Entrez Gene ID: | 9931 |
Gene Name: | helicase with zinc finger |
Gene Aliases: |
DHRC, DRHC, HUMORF5 |
Location: |
Chr.17:67070438-67245989 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HELZ | NM_014877.3 | NP_055692.2 | ||
XM_005257888.4 | XP_005257945.1 | |||
XM_005257889.4 | XP_005257946.1 | |||
XM_006722214.3 | XP_006722277.1 | |||
XM_006722215.3 | XP_006722278.1 | |||
XM_011525544.2 | XP_011523846.1 | |||
XM_017025477.1 | XP_016880966.1 | |||
XM_017025478.1 | XP_016880967.1 | |||
AA885317.1 | ||||
BC130582.1 | ||||
BC144083.1 | ||||
BP353830.1 |
Target Gene Details
Entrez Gene ID: | 101928021 |
Gene Name: | uncharacterized LOC101928021 |
Gene Aliases: |
- |
Location: |
Chr.17:67244776-67262080 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928021 | NR_136398.1 | |||
BI462408.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
