Assay Details
Target Gene Details
Entrez Gene ID: | 26033 |
Gene Name: | attractin like 1 |
Gene Aliases: |
ALP, bA338L11.1, bA454H24.1 |
Location: |
Chr.10:115093365-115948999 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATRNL1 | NM_001276282.3 | 6 | 1305 | NP_001263211.1 |
| NM_207303.4 | 6 | 1305 | NP_997186.1 | |
| NR_074088.2 | 8 | 1195 | ||
| XM_011539587.1 | 6 | 1300 | XP_011537889.1 | |
| XM_011539588.2 | 6 | 850 | XP_011537890.1 | |
| XM_011539589.2 | 6 | 1300 | XP_011537891.1 | |
| XM_011539590.2 | 6 | 3202 | XP_011537892.1 | |
| XM_011539591.2 | 6 | 1173 | XP_011537893.1 | |
| XM_017016035.1 | 6 | 1300 | XP_016871524.1 | |
| XM_017016036.1 | 6 | 1300 | XP_016871525.1 | |
| XM_017016037.1 | 5 | 560 | XP_016871526.1 | |
| XM_017016038.1 | 6 | 8512 | XP_016871527.1 | |
| AK127277.1 | 6 | 783 | BAC86914.1 | |
| AY442317.1 | 6 | 919 | AAR14297.1 | |
| BC035157.1 | 6 | 1311 | AAH35157.1 | |
| BC047716.1 | 6 | 1250 | AAH47716.1 | |
| BC139916.1 | 6 | 1294 | ||
| BC139923.1 | 6 | 1294 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831998 | Chr.10:115063088 - 115209866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map