Assay Details
Target Gene Details
Entrez Gene ID: | 124149 |
Gene Name: | ankyrin repeat domain 26 pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.16:46469337-46569097 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD26P1 | NR_026556.1 | |||
| CD000265.1 | ||||
| CR749347.1 | CAH18200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526656 | Chr.16:46505480 - 46812751 on Build GRCh38 | Gain |
|
| nsv1056162 | Chr.16:46429858 - 46621604 on Build GRCh38 | Gain |
|
| esv3571153 | Chr.16:46433586 - 46640059 on Build GRCh38 | Gain |
|
| nsv833216 | Chr.16:46467385 - 46637948 on Build GRCh38 | Loss |
|
| esv3571142 | Chr.16:46430577 - 46840772 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map