Assay Details
Target Gene Details
Entrez Gene ID: | 4809 |
Gene Name: | SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5) |
Gene Aliases: |
15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1 |
Location: |
Chr.22:41673933-41690492 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNU13 | NM_001003796.1 | 3 | 706 | NP_001003796.1 |
| NM_005008.3 | 4 | 938 | NP_004999.1 | |
| BC019282.2 | 2 | 594 | AAH19282.1 | |
| BC095439.1 | 3 | 675 | AAH95439.1 | |
| CF552526.1 | 3 | 706 | ||
| CR456531.1 | CAG30417.1 | |||
| D50420.1 | 3 | 687 | BAA23363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map