Assay Details
Target Gene Details
Entrez Gene ID: | 117196 |
Gene Name: | MAS related GPR family member X4 |
Gene Aliases: |
GPCR, MRGX4, SNSR6 |
Location: |
Chr.11:18172837-18174280 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRX4 | NM_054032.3 | 1 | 653 | NP_473373.2 |
| BC095509.1 | 1 | 663 | AAH95509.1 | |
| BC107099.1 | 1 | 233 | AAI07100.1 | |
| BC107100.1 | 1 | 233 | AAI07101.1 | |
| CN835944.1 | 1 | 800 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23532 | Chr.11:18117115 - 18179170 on Build GRCh38 | Gain |
|
| nsv518373 | Chr.11:18143140 - 18173801 on Build GRCh38 | Loss |
|
| esv2744166 | Chr.11:18129836 - 18177209 on Build GRCh38 | Deletion |
|
| nsv1038257 | Chr.11:18147863 - 18176762 on Build GRCh38 | Loss |
|
| esv2762898 | Chr.11:18145233 - 18177333 on Build GRCh38 | Gain |
|
| nsv971991 | Chr.11:18168150 - 18178928 on Build GRCh38 | Duplication |
|
| esv2668662 | Chr.11:18139517 - 18175575 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map