Assay Details
Target Gene Details
Entrez Gene ID: | 63926 |
Gene Name: | ankyrin repeat and EF-hand domain containing 1 |
Gene Aliases: |
ANKRD5 |
Location: |
Chr.20:10034999-10056764 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKEF1 | NM_001303472.1 | NP_001290401.1 | ||
| NM_022096.5 | NP_071379.3 | |||
| NM_198798.2 | NP_942093.1 | |||
| XM_005260792.2 | XP_005260849.1 | |||
| AK097689.1 | ||||
| BC022878.2 | AAH22878.1 | |||
| BI562273.1 | ||||
| BI826033.1 | ||||
| BP279314.1 |
Target Gene Details
Entrez Gene ID: | 100131208 |
Gene Name: | SNAP25 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:10023812-10219506 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAP25-AS1 | NR_040710.1 | |||
| AK124961.1 | ||||
| DA318406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833918 | Chr.20:9950381 - 10077106 on Build GRCh38 | Loss |
|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv585414 | Chr.20:10005758 - 10050283 on Build GRCh38 | Loss |
|
| nsv1063924 | Chr.20:10003340 - 10100148 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map