Assay Details
Target Gene Details
Entrez Gene ID: | 64395 |
Gene Name: | germ cell-less, spermatogenesis associated 1 |
Gene Aliases: |
BTBD13, GCL, GCL1, SPATA29 |
Location: |
Chr.2:69829606-69881396 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMCL1 | NM_178439.4 | NP_848526.1 | ||
| XM_011533033.1 | XP_011531335.1 | |||
| XM_011533034.2 | XP_011531336.1 | |||
| XM_017004704.1 | XP_016860193.1 | |||
| XM_017004705.1 | XP_016860194.1 | |||
| AK023119.1 | BAB14416.1 | |||
| AK023261.1 | BAB14494.1 | |||
| AK024042.1 | BAB14796.1 | |||
| BC007420.2 | AAH07420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834254 | Chr.2:69745440 - 69921396 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map