Assay Details
Target Gene Details
Entrez Gene ID: | 4841 |
Gene Name: | non-POU domain containing, octamer-binding |
Gene Aliases: |
MRXS34, NMT55, NRB54, P54, P54NRB, PPP1R114 |
Location: |
Chr.X:71283192-71301168 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NONO | NM_001145408.1 | NP_001138880.1 | ||
| NM_001145409.1 | NP_001138881.1 | |||
| NM_001145410.1 | NP_001138882.1 | |||
| NM_007363.4 | NP_031389.3 | |||
| AK291140.1 | ||||
| AK297144.1 | ||||
| AU119361.1 | ||||
| BC002364.1 | AAH02364.1 | |||
| BC003129.1 | AAH03129.1 | |||
| BC012141.1 | AAH12141.1 | |||
| BC028299.1 | AAH28299.1 | |||
| BC069616.1 | AAH69616.1 | |||
| BC069639.1 | AAH69639.1 | |||
| CR456761.1 | CAG33042.1 | |||
| KU178234.1 | ||||
| KU178235.1 | ||||
| L14599.1 | AAC37578.1 | |||
| U02493.1 | AAA03427.1 | |||
| U89867.1 | AAC51852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map