Assay Details
Target Gene Details
Entrez Gene ID: | 54662 |
Gene Name: | TBC1 domain family member 13 |
Gene Aliases: |
- |
Location: |
Chr.9:128787207-128810432 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D13 | NM_001286772.1 | NP_001273701.1 | ||
| NM_018201.4 | NP_060671.3 | |||
| XM_005252060.2 | XP_005252117.1 | |||
| AB209565.1 | BAD92802.1 | |||
| AB449899.1 | ||||
| AK001605.1 | BAA91784.1 | |||
| AK123828.1 | ||||
| AK289777.1 | ||||
| AK295183.1 | ||||
| AK307556.1 | ||||
| BC132817.1 | ||||
| BC136509.1 | ||||
| BC150311.1 | ||||
| BG716911.1 | ||||
| DC325060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615412 | Chr.9:128754559 - 128824961 on Build GRCh38 | Loss |
|
| nsv615411 | Chr.9:128727172 - 128829871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map