Assay Details
Target Gene Details
Entrez Gene ID: | 3400 |
Gene Name: | inhibitor of DNA binding 4, HLH protein |
Gene Aliases: |
IDB4, bHLHb27 |
Location: |
Chr.6:19837370-19842200 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID4 | NM_001546.3 | 3 | 1640 | NP_001537.1 |
| AK130851.1 | ||||
| BC014941.1 | 3 | 1624 | AAH14941.1 |
More Information
Set Membership: |
|

Genomic Map