Assay Details
Target Gene Details
Entrez Gene ID: | 9917 |
Gene Name: | family with sequence similarity 20 member B |
Gene Aliases: |
gxk1 |
Location: |
Chr.1:179025804-179076574 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM20B | NM_001324310.1 | NP_001311239.1 | ||
| NM_001324311.1 | NP_001311240.1 | |||
| NM_014864.3 | NP_055679.1 | |||
| XM_017003001.1 | XP_016858490.1 | |||
| XM_017003002.1 | XP_016858491.1 | |||
| AB007944.1 | ||||
| AB480690.1 | ||||
| AK289989.1 | ||||
| BC046441.1 | AAH46441.1 | |||
| BC051794.1 | AAH51794.1 |
More Information
Set Membership: |
|

Genomic Map