Assay Details
Target Gene Details
Entrez Gene ID: | 7569 |
Gene Name: | zinc finger protein 182 |
Gene Aliases: |
HHZ150, KOX14, ZNF21, Zfp182 |
Location: |
Chr.X:47974851-48003995 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF182 | NM_001007088.1 | 6 | 2393 | NP_001007089.1 |
| NM_001178099.1 | 7 | 2509 | NP_001171570.1 | |
| NM_006962.1 | 7 | 2487 | NP_008893.1 | |
| AK097687.1 | 1 | 1046 | ||
| AK122874.1 | 7 | 2492 | ||
| AK130362.1 | 1 | 1613 | ||
| BC106875.2 | 6 | 2357 | AAI06876.1 | |
| BC114216.1 | 6 | 2370 | AAI14217.1 | |
| BX537686.1 | 1 | 1378 | ||
| BX648401.1 | 1 | 1709 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758866 | Chr.X:47907511 - 48287275 on Build GRCh38 | Gain+Loss |
|
| nsv6890 | Chr.X:47968293 - 48168190 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map