Assay Details
Target Gene Details
Entrez Gene ID: | 8934 |
Gene Name: | RAB29, member RAS oncogene family |
Gene Aliases: |
RAB7L, RAB7L1 |
Location: |
Chr.1:205767986-205775487 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB29 | NM_001135662.1 | 6 | 1192 | NP_001129134.1 |
| NM_001135663.1 | 4 | 1407 | NP_001129135.1 | |
| NM_001135664.1 | 5 | 1039 | NP_001129136.1 | |
| NM_003929.2 | 6 | 1293 | NP_003920.1 | |
| XM_005245569.1 | 6 | 1191 | XP_005245626.1 | |
| XM_005245570.1 | 6 | 1244 | XP_005245627.1 | |
| XM_005245571.1 | 6 | 1169 | XP_005245628.1 | |
| XM_006711605.3 | 5 | 935 | XP_006711668.1 | |
| XM_006711606.2 | 5 | 899 | XP_006711669.1 | |
| XM_017002748.1 | 5 | 1194 | XP_016858237.1 | |
| XM_017002749.1 | 5 | 1172 | XP_016858238.1 | |
| XM_017002750.1 | 5 | 1104 | XP_016858239.1 | |
| AK303879.1 | 4 | 1158 | ||
| AK308359.1 | 5 | 1008 | ||
| AK311085.1 | 4 | 916 | ||
| BC002585.2 | 6 | 1126 | AAH02585.1 | |
| BX647960.1 | 6 | 1258 | ||
| D84488.1 | 5 | 993 | BAA22160.1 |
More Information
Set Membership: |
|

Genomic Map