Assay Details
Target Gene Details
Entrez Gene ID: | 23174 |
Gene Name: | zinc finger CCHC-type containing 14 |
Gene Aliases: |
BDG-29, BDG29 |
Location: |
Chr.16:87406246-87492948 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZCCHC14 | NM_015144.2 | 13 | 5959 | NP_055959.1 |
| AB011151.2 | 13 | 5817 | BAA25505.2 | |
| AB030243.1 | BAB83129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv499n67 | Chr.16:87353978 - 87409229 on Build GRCh38 | Gain |
|
| nsv833320 | Chr.16:87315844 - 87501226 on Build GRCh38 | Gain |
|
| nsv428330 | Chr.16:87336922 - 87633082 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map