Assay Details
Target Gene Details
Entrez Gene ID: | 10785 |
Gene Name: | WD repeat domain 4 |
Gene Aliases: |
TRM82, TRMT82 |
Location: |
Chr.21:42843075-42879865 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR4 | NM_001260474.1 | NP_001247403.1 | ||
| NM_001260475.1 | NP_001247404.1 | |||
| NM_001260476.1 | NP_001247405.1 | |||
| NM_001260477.1 | NP_001247406.1 | |||
| NM_018669.5 | NP_061139.2 | |||
| NM_033661.4 | NP_387510.1 | |||
| NR_048535.1 | ||||
| XM_011529433.1 | XP_011527735.1 | |||
| XM_017028262.1 | XP_016883751.1 | |||
| XM_017028263.1 | XP_016883752.1 | |||
| XM_017028264.1 | XP_016883753.1 | |||
| AJ243912.1 | CAB93144.1 | |||
| AJ243913.1 | CAB93145.1 | |||
| AK056343.1 | ||||
| AK292923.1 | ||||
| AK298015.1 | ||||
| AK315008.1 | ||||
| BC001074.2 | AAH01074.1 | |||
| BC006341.2 | AAH06341.1 | |||
| CD368164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587657 | Chr.21:42752699 - 43087910 on Build GRCh38 | Gain |
|
| esv3584478 | Chr.21:42827914 - 42993457 on Build GRCh38 | Gain |
|
| esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion |
|
| nsv587659 | Chr.21:42833569 - 42853748 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map