Assay Details
Target Gene Details
Entrez Gene ID: | 23270 |
Gene Name: | TSPY like 4 |
Gene Aliases: |
dJ486I3.2 |
Location: |
Chr.6:116249964-116254098 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPYL4 | NM_021648.4 | 1 | 200 | NP_067680.3 |
| AB018264.1 | 1 | 169 | ||
| AK091559.1 | ||||
| AK302542.1 | 1 | 203 | ||
| AL134793.1 | 1 | 200 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576211 | Chr.6:116095308 - 116431057 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map