Assay Details
Target Gene Details
Entrez Gene ID: | 10266 |
Gene Name: | receptor activity modifying protein 2 |
Gene Aliases: |
- |
Location: |
Chr.17:42761194-42763041 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAMP2 | NM_005854.2 | NP_005845.2 | ||
| AJ001015.1 | CAA04473.1 | |||
| AL548945.3 | ||||
| AY265458.1 | AAP23299.1 | |||
| BC027975.1 | AAH27975.1 | |||
| EF687002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map