Assay Details
Target Gene Details
Entrez Gene ID: | 84844 |
Gene Name: | PHD finger protein 5A |
Gene Aliases: |
INI, Rds3, SAP14b, SF3B7, SF3b14b, bK223H9.2 |
Location: |
Chr.22:41459717-41468704 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF5A | NM_032758.3 | NP_116147.1 | ||
| BC007321.2 | AAH07321.1 | |||
| BC075808.1 | AAH75808.1 | |||
| BU584712.1 | ||||
| CR456398.1 | CAG30284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
|
| nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map