Assay Details
Target Gene Details
Entrez Gene ID: | 6787 |
Gene Name: | NIMA related kinase 4 |
Gene Aliases: |
NRK2, STK2, pp12301 |
Location: |
Chr.3:52707936-52770949 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK4 | NM_001193533.1 | 15 | 2563 | NP_001180462.1 |
| NM_003157.4 | 16 | 2830 | NP_003148.2 | |
| XM_006713310.2 | 15 | 2689 | XP_006713373.1 | |
| AK294165.1 | 15 | 2470 | ||
| EF560744.1 | ||||
| L20321.1 | 16 | 2805 | AAA36658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508219 | Chr.3:52707177 - 52769883 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map