Assay Details
Target Gene Details
Entrez Gene ID: | 53373 |
Gene Name: | two pore segment channel 1 |
Gene Aliases: |
TPC1 |
Location: |
Chr.12:113221429-113298589 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPCN1 | NM_001143819.2 | NP_001137291.1 | ||
| NM_017901.5 | NP_060371.2 | |||
| XM_011538490.2 | XP_011536792.1 | |||
| XM_011538492.2 | XP_011536794.1 | |||
| XM_011538493.2 | XP_011536795.1 | |||
| XM_017019480.1 | XP_016874969.1 | |||
| XM_017019481.1 | XP_016874970.1 | |||
| XM_017019482.1 | XP_016874971.1 | |||
| AB032995.2 | BAA86483.2 | |||
| AK057414.1 | ||||
| AK093490.1 | ||||
| AY083666.1 | AAM01199.1 | |||
| BC136795.1 | ||||
| BC136796.1 | ||||
| BC142664.1 | ||||
| BC150203.1 | ||||
| BC152423.1 | ||||
| DA777641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map