Assay Details
Target Gene Details
Entrez Gene ID: | 8407 |
Gene Name: | transgelin 2 |
Gene Aliases: |
HA1756 |
Location: |
Chr.1:159918107-159925542 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAGLN2 | NM_001277223.1 | NP_001264152.1 | ||
| NM_001277224.1 | NP_001264153.1 | |||
| NM_003564.2 | NP_003555.1 | |||
| AK291562.1 | ||||
| AY007127.1 | AAG01993.1 | |||
| BC002616.1 | AAH02616.1 | |||
| BC009357.2 | AAH09357.1 | |||
| CD051174.1 | ||||
| CF128621.1 | ||||
| CR542126.1 | CAG46923.1 | |||
| CR542136.1 | CAG46933.1 | |||
| D21261.1 | ||||
| GU727636.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464150 | Chr.1:159891127 - 159938603 on Build GRCh38 | Gain |
|
| nsv548053 | Chr.1:159915710 - 159938603 on Build GRCh38 | Loss |
|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
| nsv946451 | Chr.1:159918291 - 159920328 on Build GRCh38 | Duplication |
|
| nsv524006 | Chr.1:159916632 - 159953321 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map