Assay Details
Target Gene Details
Entrez Gene ID: | 347168 |
Gene Name: | olfactory receptor family 1 subfamily J member 1 |
Gene Aliases: |
OR9-18, hg32 |
Location: |
Chr.9:122476958-122477926 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1J1 | NM_001004451.1 | NP_001004451.1 | ||
| BC136933.1 | 1 | 36 | ||
| BC136938.1 | 1 | 36 |
Target Gene Details
Entrez Gene ID: | 26740 |
Gene Name: | olfactory receptor family 1 subfamily J member 2 |
Gene Aliases: |
HG152, HSA5, OR1J3, OR1J5, OR9-19, OST044 |
Location: |
Chr.9:122403407-122514097 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1J2 | XM_017014623.1 | XP_016870112.1 | ||
| AK123016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6704 | Chr.9:122461748 - 122506664 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138102077] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map