Assay Details
Target Gene Details
Entrez Gene ID: | 348 |
Gene Name: | apolipoprotein E |
Gene Aliases: |
AD2, APO-E, LDLCQ5, LPG |
Location: |
Chr.19:44905749-44909395 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOE | NM_000041.3 | NP_000032.1 | ||
| NM_001302688.1 | NP_001289617.1 | |||
| NM_001302689.1 | NP_001289618.1 | |||
| NM_001302690.1 | NP_001289619.1 | |||
| NM_001302691.1 | NP_001289620.1 | |||
| AK308546.1 | ||||
| AK314898.1 | ||||
| BC003557.1 | AAH03557.1 | |||
| BI550631.1 | ||||
| BM042032.1 | ||||
| CA771535.1 | 1 | 275 | ||
| DB486513.1 | ||||
| HY056112.1 | ||||
| HY074384.1 | ||||
| HY074888.1 | ||||
| K00396.1 | AAB59546.1 | |||
| KU177910.1 | ||||
| KU177911.1 | ||||
| KU177912.1 | ||||
| KU177913.1 | ||||
| M12529.1 | AAB59518.1 | |||
| Z70760.1 | CAA94806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509747 | Chr.19:44877936 - 44943426 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map