Assay Details
Target Gene Details
Entrez Gene ID: | 64081 |
Gene Name: | phenazine biosynthesis like protein domain containing |
Gene Aliases: |
HEL-S-306, MAWBP, MAWDBP |
Location: |
Chr.10:68282660-68332958 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBLD | NM_001033083.1 | 9 | 960 | NP_001028255.1 |
| NM_022129.3 | NP_071412.2 | |||
| XM_005270028.3 | XP_005270085.1 | |||
| XM_011540060.2 | XP_011538362.1 | |||
| XM_017016513.1 | XP_016872002.1 | |||
| AB049758.1 | BAB16606.1 | |||
| AI114748.1 | ||||
| AI439144.1 | 1 | 270 | ||
| AK027673.1 | BAB55285.1 | |||
| AK312275.1 | ||||
| BC009738.1 | AAH09738.1 | |||
| GQ472233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623705 | Chr.10:68099008 - 68297411 on Build GRCh38 | Gain |
|
| esv3623707 | Chr.10:68214112 - 68304911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map